Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jakmip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178394.4 | NP_848481.2 | 5 | 1137 | ||
GenBank mRNA | AK017977.1 | - | 5 | 1092 | ||
AK077298.1 | - | 5 | 1079 | |||
BC138646.1 | - | 4 | 989 | 8861666 | ||
BC138649.1 | - | 4 | 989 | 8861669 |