Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029509.4 | NP_083785.3 | 507 | |||
GenBank mRNA | BC031475.1 | - | 502 | 2654170 | ||
DQ295175.1 | - | 3, 4 | 422 |