Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Las1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152822.3 | NP_690035.2 | 5 | 809 | ||
GenBank mRNA | AK089160.2 | - | 4 | 2600 | ||
AK163919.1 | - | 4 | 2636 | |||
BC004735.1 | - | 4 | 401 | |||
BC032218.1 | - | 5 | 732 | |||
BC141155.1 | - | 5 | 746 | 9055767 | ||
BQ573371.1 | - | 4 | 389 |