Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001503.1 | NP_001001503.1 | 3 | 313 | ||
NM_021075.3 | NP_066553.3 | 4 | 1408 | |||
XM_011529586.2 | XP_011527888.1 | 5 | 1468 | |||
XM_017028359.1 | XP_016883848.1 | 4 | 371 | |||
GenBank mRNA | AK056041.1 | - | 6 | 1865 | ||
AK289586.1 | - | 3 | 267 | |||
AK308228.1 | - | 4 | 1365 | |||
AY173949.1 | - | 3 | 271 | |||
BC021217.2 | - | 4 | 1395 | 4302265 | ||
BC033766.1 | - | 3 | 271 | |||
BC054016.1 | - | 3 | 314 | |||
CR542174.1 | - | 3 | 247 | |||
CR542190.1 | - | 3 | 247 |