Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam131b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113327.2 | NP_001106798.1 | 3 | 334 | ||
NM_001286584.1 | NP_001273513.1 | 4 | 310 | |||
NM_029528.5 | NP_083804.3 | 4 | 370 | |||
GenBank mRNA | AK018206.1 | - | 4 | 309 | ||
AK018325.1 | - | 4 | 316 | |||
AK047763.1 | - | 4 | 296 | |||
AK158869.1 | - | 4 | 263 | |||
AK163654.1 | - | 3 | 255 | |||
BC115595.1 | - | 3 | 100 | |||
BC115596.1 | - | 3 | 100 | |||
BY260739.1 | - | 3 | 335 |