Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198006.4 | NP_932123.3 | 2 | 273 | ||
GenBank mRNA | AK018255.1 | - | 2 | 149 | ||
AK050432.1 | - | 2 | 179 | |||
AK166651.1 | - | 2 | 203 | |||
AK177444.1 | - | 2 | 94 | |||
AK218336.1 | - | 2 | 94 | |||
BB860009.1 | - | 2 | 224 | |||
BB863739.1 | - | 2 | 216 | |||
BC002137.1 | - | 2 | 180 | 3484305 | ||
BY779699.1 | - | 2 | 273 |