Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175235.3 | NP_780444.2 | 10, 11 | 1291 | ||
GenBank mRNA | AK018246.1 | - | 9, 10 | 1079 | ||
AK034216.1 | - | 9, 10 | 1087 | |||
BC052406.1 | - | 10, 11 | 1291 | 5716119 | ||
BC057083.1 | - | 9, 10 | 1073 | 5707546 |