Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med13l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172424.4 | NP_766012.3 | 28 | 6161 | ||
GenBank mRNA | AK122419.1 | - | 19 | 4449 | ||
AK154895.1 | - | 28 | 6168 | |||
AK162842.1 | - | 5 | 559 | |||
AK170157.1 | - | 7 | 1050 | |||
AY338464.1 | - | 28 | 6138 | |||
BC040283.1 | - | 8 | 2652 | |||
BC049985.1 | - | 6 | 1387 | |||
BC052320.1 | - | 12 | 2848 | |||
BC072653.1 | - | 21 | 5111 |