Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029536.3 | NP_083812.1 | 2 | 1062 | ||
GenBank mRNA | AK018327.1 | - | 2 | 1090 | ||
AK034479.1 | - | 2 | 1006 | |||
AK038566.1 | - | 2 | 334 | |||
AK042723.1 | - | 2 | 1039 | |||
AK045935.1 | - | 2 | 1018 | |||
BC113184.1 | - | 2 | 913 | 40061761 | ||
BC113791.1 | - | 2 | 913 | 40061764 |
Set Membership: |
G-protein coupled receptor |