Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029536.3 | NP_083812.1 | 3 | 1222 | ||
GenBank mRNA | AK018327.1 | - | 3 | 1250 | ||
AK034479.1 | - | 3 | 1166 | |||
AK038566.1 | - | 3 | 494 | |||
AK042723.1 | - | 3 | 1199 | |||
BC113184.1 | - | 3 | 1073 | 40061761 | ||
BC113791.1 | - | 3 | 1073 | 40061764 |
Set Membership: |
G-protein coupled receptor |