Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029557.1 | NP_083833.1 | 7 | 581 | ||
GenBank mRNA | AK002758.1 | - | 7 | 566 | ||
AK089005.1 | - | 7 | 548 | |||
BC126953.1 | - | 6 | 705 |