Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033174.1 | NP_001028346.1 | 3 | 770 | ||
GenBank mRNA | BC107327.1 | - | 3 | 504 | ||
BC107328.2 | - | 3 | 504 |