Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ropn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030744.2 | NP_109669.1 | 5 | 630 | ||
GenBank mRNA | AF178531.1 | - | 4 | 553 | ||
AK160142.1 | - | 3 | 372 | |||
BC049544.1 | - | 5 | 630 | 6740976 |