Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110002H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029623.2 | NP_083899.1 | 16 | 1520 | ||
GenBank mRNA | AF003352.1 | - | 4 | 406 | ||
AK013979.1 | - | 10 | 906 | |||
AK144623.1 | - | 16 | 1521 | |||
AK154240.1 | - | 16 | 1515 | |||
AK159691.1 | - | 16 | 1515 | |||
BC012285.1 | - | 15 | 1683 | |||
BC021846.1 | - | 16 | 1503 | 5027035 | ||
BC044861.1 | - | 1 | 3161 |