Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110002H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029623.2 | NP_083899.1 | 14 | 1311 | ||
GenBank mRNA | AF003352.1 | - | 2 | 197 | ||
AK144623.1 | - | 14 | 1312 | |||
AK154240.1 | - | 14 | 1306 | |||
AK159691.1 | - | 14 | 1306 | |||
AK182921.1 | - | 5 | 360 | |||
BC012285.1 | - | 13 | 1474 | |||
BC021846.1 | - | 14 | 1294 | 5027035 | ||
BC044861.1 | - | 1 | 270 |