Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006158.4 | NP_006149.2 | 2 | 1457 | ||
GenBank mRNA | AK057731.1 | - | 3 | 1031 | ||
AK075003.1 | - | 2 | 1200 | |||
AK127430.1 | - | 4 | 900 | |||
AK225975.1 | - | 2 | 1200 | |||
AK299095.1 | - | 3 | 1042 | |||
AK314591.1 | - | 2 | 1200 | |||
AL713644.1 | - | 3 | 633 | |||
AY156690.1 | - | 2 | 1103 | |||
BC039237.1 | - | 2 | 1200 | 4280159 | ||
KU178228.1 | - | 3 | 824 | |||
KU178229.1 | - | 2 | 1103 |