Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164641.1 | NP_001158113.1 | 6 | 1160 | ||
NM_001164642.1 | NP_001158114.1 | 5 | 1133 | |||
NM_001164643.1 | NP_001158115.1 | 5 | 1133 | |||
NM_029640.2 | NP_083916.1 | 5 | 388 | |||
NM_180662.2 | NP_850993.2 | 5 | 1133 | |||
GenBank mRNA | AK028504.1 | - | 5 | 975 | ||
AK029106.1 | - | 6 | 1002 | |||
AK031788.1 | - | 5 | 981 | |||
AK138682.1 | - | 5 | 980 | |||
AK156739.1 | - | 5 | 958 | |||
AK214984.1 | - | 2 | 104 | |||
AY630620.1 | - | 5 | 386 | |||
BC034590.1 | - | 5 | 386 | 5364921 | ||
BC070463.1 | - | 5 | 1133 | 30613261 |