Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164641.1 | NP_001158113.1 | 13, 14 | 2233 | ||
NM_001164642.1 | NP_001158114.1 | 12, 13 | 2206 | |||
NM_029640.2 | NP_083916.1 | 12, 13 | 1461 | |||
NM_180662.2 | NP_850993.2 | 12, 13 | 2206 | |||
GenBank mRNA | AK028504.1 | - | 12, 13 | 2048 | ||
AK029106.1 | - | 13, 14 | 2075 | |||
AK138682.1 | - | 12, 13 | 2053 | |||
AK156739.1 | - | 13, 14 | 2031 | |||
AK173287.1 | - | 2, 3 | 205 | |||
AY630620.1 | - | 12, 13 | 1459 | |||
BC034590.1 | - | 12, 13 | 1459 | 5364921 | ||
BC038938.1 | - | 6, 7 | 776 | |||
BC070463.1 | - | 12, 13 | 2206 | 30613261 |