Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 7 | 852 | ||
NM_002498.2 | NP_002489.1 | 7 | 852 | |||
NM_152720.2 | NP_689933.1 | 7 | 871 | |||
NR_027415.2 | 7 | 852 | ||||
GenBank mRNA | AB072828.1 | - | 6 | 521 | ||
AK131359.1 | - | 6 | 1770 | |||
AK290259.1 | - | 7 | 584 | |||
AL833951.1 | - | 7 | 711 | |||
BC019916.1 | - | 7 | 579 | 5000918 | ||
BQ648157.1 | - | 7 | 787 | |||
CV029372.1 | - | 6 | 476 | |||
Z25434.1 | - | 4 | 251 | |||
Z29067.1 | - | 5 | 335 |