Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hectd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175244.3 | NP_780453.1 | 8 | 1264 | ||
GenBank mRNA | AK051385.1 | - | 8 | 1277 | ||
AK139305.1 | - | 8 | 1292 | |||
AK154378.1 | - | 8 | 1296 | |||
AK155290.1 | - | 8 | 1277 | |||
AK155676.1 | - | 8 | 1277 | |||
AK163323.1 | - | 8 | 1299 | |||
AK165482.1 | - | 8 | 1296 | |||
AK170930.1 | - | 8 | 1277 | |||
BC070411.1 | - | 8 | 1264 | 5706209 |