Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054043.3 | NP_473384.1 | 11, 12 | 932 | ||
GenBank mRNA | AK007122.1 | - | 1, 2 | 141 | ||
AK136156.1 | - | 7, 8 | 740 | |||
AK153928.1 | - | 11, 12 | 805 | |||
AK169846.1 | - | 11, 12 | 805 |