Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stambpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029682.4 | NP_083958.3 | 6 | 873 | ||
GenBank mRNA | AB010121.1 | - | 6 | 568 | ||
AB010122.1 | - | 6 | 532 | |||
AB066211.1 | - | 6 | 873 | |||
AB066212.1 | - | 2 | 222 | |||
AB066213.1 | - | 6 | 556 | |||
AK007087.1 | - | 2 | 206 | |||
AK085665.1 | - | 5 | 3264 | |||
BC018343.1 | - | 6 | 599 | 4010423 | ||
BC023097.1 | - | 6 | 842 |