Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029686.4 | NP_083962.4 | 35, 36 | 6037 | ||
GenBank mRNA | AK131811.1 | - | 4, 5 | 729 | ||
AY164484.1 | - | 35, 36 | 6040 |