Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029696.4 | NP_083972.3 | 5 | 828 | ||
GenBank mRNA | BC050786.1 | - | 5 | 828 | 6703279 |