Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293300.1 | NP_001280229.1 | 7 | 1329 | ||
NM_001293301.1 | NP_001280230.1 | 7 | 1329 | |||
NM_029709.2 | NP_083985.2 | 7 | 1329 | |||
GenBank mRNA | AJ276962.1 | - | 7 | 929 | ||
AK031181.1 | - | 7 | 931 | |||
AK042402.1 | - | 7 | 1333 | |||
AK080782.1 | - | 7 | 1045 | |||
BC094432.1 | - | 6 | 870 | 30674963 | ||
BC157938.1 | - | 7 | 931 | |||
BC158064.1 | - | 7 | 932 | 9007363 |