Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029714.3 | NP_083990.2 | 2 | 186 | ||
GenBank mRNA | AK006911.1 | - | 2 | 177 | ||
AK077059.1 | - | 2 | 168 | |||
AK138347.1 | - | 2 | 186 | |||
BC144969.1 | - | 2 | 165 | |||
BY724013.1 | - | 2 | 186 | |||
GQ225581.1 | - | 1 | 87 |
Gene Symbol: Catsperg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164658.1 | NP_001158130.1 | 2 | 216 | ||
GenBank mRNA | AK039202.1 | - | 2 | 221 | ||
AK042364.1 | - | 2 | 148 | |||
BB633050.1 | - | 2 | 216 |