Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082484.2 | NP_001075953.1 | 5 | 1026 | ||
NM_029721.2 | NP_083997.1 | 5 | 1026 | |||
GenBank mRNA | AK017836.1 | - | 2 | 97 | ||
AK122296.1 | - | 5 | 838 | |||
AK147452.1 | - | 5 | 1013 | |||
BC053495.1 | - | 5 | 838 |