Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsdma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029727.2 | NP_084003.2 | 11 | 1234 | ||
GenBank mRNA | AK008613.1 | - | 9 | 887 | ||
AK008663.1 | - | 11 | 1230 | |||
BC109335.1 | - | 10 | 1137 | 40052139 | ||
BC109336.1 | - | 11 | 1168 | 40052146 |