Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290523.1 | NP_001277452.1 | 4 | 451 | ||
NM_001290524.1 | NP_001277453.1 | 4 | 451 | |||
NM_029730.4 | NP_084006.2 | 4 | 451 | |||
GenBank mRNA | AK010480.1 | - | 4 | 373 | ||
AK031405.1 | - | 4 | 376 | |||
AK035985.1 | - | 4 | 411 | |||
AK039893.1 | - | 4 | 402 | |||
AK135188.1 | - | 4 | 406 | |||
AK156572.1 | - | 4 | 394 | |||
BC026425.1 | - | 4 | 377 | 4223175 |