Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mospd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290523.1 | NP_001277452.1 | 5 | 490 | ||
NM_001290524.1 | NP_001277453.1 | 5 | 490 | |||
NM_029730.4 | NP_084006.2 | 5 | 490 | |||
GenBank mRNA | AK010480.1 | - | 5 | 412 | ||
AK031405.1 | - | 5 | 415 | |||
AK035985.1 | - | 5 | 450 | |||
AK039893.1 | - | 5 | 441 | |||
AK135188.1 | - | 5 | 445 | |||
AK156572.1 | - | 5 | 433 | |||
BC026425.1 | - | 5 | 416 | 4223175 |