Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029742.2 | NP_084018.1 | 6 | 931 | ||
GenBank mRNA | AK010783.1 | - | 6 | 796 | ||
AK039732.1 | - | 5 | 833 | |||
AK050289.1 | - | 4 | 552 | |||
AK053834.1 | - | 5 | 803 | |||
AK085116.1 | - | 5 | 882 | |||
AK085130.1 | - | 6 | 889 | |||
AK172923.1 | - | 5 | 844 | |||
BC057064.1 | - | 2 | 882 | |||
BC060132.1 | - | 6 | 931 | 6409751 |