Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bri3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029752.2 | NP_084028.1 | 3 | 526 | ||
GenBank mRNA | AK010810.1 | - | 3 | 469 | ||
AK043288.1 | - | 3 | 468 | |||
AK044565.1 | - | 3 | 414 | |||
AK143538.1 | - | 3 | 485 | |||
BB618776.1 | - | 3 | 516 | |||
BC039941.1 | - | 3 | 484 | 4011957 |