Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278669.1 | NP_001265598.1 | 7 | 2383 | ||
NM_001278670.1 | NP_001265599.1 | 7 | 2383 | |||
NM_001278672.1 | NP_001265601.1 | 7 | 2009 | |||
NM_001278673.1 | NP_001265602.1 | 6 | 910 | |||
NM_001278675.1 | NP_001265604.1 | 7 | 2009 | |||
NM_006162.4 | NP_006153.2 | 7 | 2383 | |||
NM_172387.2 | NP_765975.1 | 7 | 2009 | |||
NM_172388.2 | NP_765976.1 | 6 | 1284 | |||
NM_172389.2 | NP_765977.1 | 7 | 2009 | |||
NM_172390.2 | NP_765978.1 | 7 | 2383 | |||
GenBank mRNA | AK292641.1 | - | 7 | 2225 | ||
AK293760.1 | - | 6 | 855 | |||
BC104753.1 | - | 7 | 2024 | 8143756 | ||
BC112243.1 | - | 7 | 2024 | 8327711 | ||
BC143457.1 | - | 7 | 1939 | |||
EU887559.1 | - | 7 | 1930 | |||
EU887560.1 | - | 7 | 1891 | |||
EU887561.1 | - | 7 | 1930 | |||
EU887562.1 | - | 7 | 1891 | |||
EU887563.1 | - | 7 | 1930 | |||
EU887564.1 | - | 7 | 1891 | |||
EU887565.1 | - | 7 | 1930 | |||
EU887566.1 | - | 7 | 1891 | |||
U08015.1 | - | 7 | 2169 | |||
U59736.1 | - | 7 | 1966 | |||
U80917.1 | - | 7 | 2299 | |||
U80918.1 | - | 7 | 1953 | |||
U80919.1 | - | 6 | 1112 |