Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ago4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153177.3 | NP_694817.2 | 3, 4 | 562 | ||
GenBank mRNA | AB081474.1 | - | 3, 4 | 307 | ||
AK030018.1 | - | 3, 4 | 578 | |||
BB616446.1 | - | 3, 4 | 562 | |||
BC096023.1 | - | 3, 4 | 344 | 30607807 |