Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002894.2 | NP_001002894.2 | 6 | 2153 | ||
GenBank mRNA | AK014932.1 | - | 2 | 311 | ||
AY596199.1 | - | 5 | 2029 | |||
AY673647.1 | - | 6 | 2153 | |||
BC141199.1 | - | 6 | 2137 | 9055811 |