Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029781.3 | NP_084057.1 | 5 | 552 | ||
GenBank mRNA | AB232632.1 | - | 4 | 307 | ||
AK018269.1 | - | 5 | 571 | |||
AK038445.1 | - | 5 | 553 | |||
AK043946.1 | - | 5 | 576 | |||
AK046113.1 | - | 5 | 361 | |||
AK142025.1 | - | 4 | 924 | |||
AK142415.1 | - | 4 | 1534 | |||
AK163455.1 | - | 5 | 560 | |||
AK200285.1 | - | 2 | 107 | |||
BC119094.1 | - | 5 | 540 | 8733843 | ||
BC119098.1 | - | 5 | 527 | 8733847 |