Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam81a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029784.2 | NP_084060.1 | 7 | 1087 | ||
GenBank mRNA | AK018282.1 | - | 7 | 1044 | ||
AK038532.1 | - | 7 | 1046 | |||
AK044090.1 | - | 7 | 1087 | |||
AK046408.1 | - | 7 | 980 | |||
AK079541.1 | - | 7 | 1047 | |||
AK135091.1 | - | 6 | 802 | |||
BC025646.1 | - | 7 | 1113 | 5324986 |