Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039179.2 | NP_001034268.1 | 4 | 927 | ||
NM_001039180.2 | NP_001034269.1 | 5 | 823 | |||
NM_029791.4 | NP_084067.1 | 4 | 927 | |||
GenBank mRNA | AJ250106.1 | - | 4 | 886 | ||
AK002683.1 | - | 4 | 609 | |||
AK122348.1 | - | 4 | 900 | |||
AK156407.1 | - | 4 | 882 | |||
AK171300.1 | - | 5 | 781 | |||
BC032198.1 | - | 4 | 884 | 5324113 |