Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133772.2 | NP_598533.2 | 10, 11 | 816 | ||
GenBank mRNA | AK004835.1 | - | 10, 11 | 929 | ||
AK154336.1 | - | 10, 11 | 817 | |||
BC002150.1 | - | 10, 11 | 775 | 3484896 | ||
CX237617.1 | - | 9, 10 | 598 |