Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029796.2 | NP_084072.1 | 2 | 169 | ||
GenBank mRNA | AB055885.1 | - | 2 | 129 | ||
AF403429.1 | - | 2 | 112 | |||
AK004940.1 | - | 1 | 117 | |||
AK009555.1 | - | 1 | 125 | |||
BC030733.1 | - | 2 | 169 | 5123909 | ||
BC092077.1 | - | 2 | 114 | 4975749 |