Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164363.1 | NP_001157835.1 | 8 | 1087 | ||
NM_001164365.1 | NP_001157837.1 | 8 | 744 | |||
NM_029810.4 | NP_084086.3 | 8 | 675 | |||
GenBank mRNA | AK008045.1 | - | 8 | 622 | ||
AK030786.1 | - | 9 | 692 | |||
AK036730.1 | - | 8 | 633 | |||
AK038458.1 | - | 5 | 380 | |||
AK132384.1 | - | 8 | 633 | |||
AK147901.1 | - | 8 | 633 | |||
AK162385.1 | - | 8 | 1052 | |||
AK169616.1 | - | 8 | 633 | |||
AK182698.1 | - | 6 | 419 | |||
BC064760.1 | - | 7 | 609 | 30436992 |