Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023850.2 | NP_076339.1 | 3 | 897 | ||
GenBank mRNA | AF280087.1 | - | 2 | 385 | ||
AK134997.1 | - | 3 | 771 | |||
BC030667.1 | - | 2 | 376 | 1245270 |