Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2ql1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145162.1 | NP_001138634.1 | 1 | 1064 | ||
GenBank mRNA | BC116979.1 | - | 1 | 114 | ||
BC116981.1 | - | 1 | 114 |