Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039201.1 | NP_001034290.1 | 3 | 357 | ||
NM_001039202.1 | NP_001034291.1 | 3 | 318 | |||
NM_029826.2 | NP_084102.1 | 3 | 318 | |||
GenBank mRNA | AK002837.1 | - | 3 | 306 | ||
AK014208.1 | - | 3 | 312 | |||
AK137285.1 | - | 3 | 310 | |||
AK147794.1 | - | 339 | ||||
AK151285.1 | - | 3 | 306 | |||
AK152162.1 | - | 3 | 306 | |||
AK152259.1 | - | 3 | 306 | |||
BC058177.1 | - | 3 | 292 | 5250739 | ||
BY069367.1 | - | 3 | 328 |