Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039201.1 | NP_001034290.1 | 4 | 488 | ||
NM_001039202.1 | NP_001034291.1 | 4 | 449 | |||
NM_029826.2 | NP_084102.1 | 4 | 449 | |||
GenBank mRNA | AK002837.1 | - | 4 | 437 | ||
AK014208.1 | - | 4 | 443 | |||
AK137285.1 | - | 4 | 441 | |||
AK147794.1 | - | 470 | ||||
AK151285.1 | - | 4 | 437 | |||
AK152162.1 | - | 4 | 437 | |||
AK152259.1 | - | 4 | 437 | |||
BC058177.1 | - | 4 | 423 | 5250739 |