Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029835.1 | NP_084111.1 | 20 | 3513 | ||
GenBank mRNA | AK051639.1 | - | 20 | 3476 | ||
AK054492.1 | - | 20 | 3480 | |||
BC158103.1 | - | 20 | 3477 | 9088180 | ||
CN537331.1 | - | 1 | 51 |