Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700109H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029843.2 | NP_084119.1 | 4, 5 | 510 | ||
GenBank mRNA | AK007150.1 | - | 4, 5 | 553 | ||
BC048606.1 | - | 4, 5 | 612 | |||
BC099581.1 | - | 4, 5 | 510 | 30920581 |