Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205391.1 | NP_001192320.1 | 3 | 428 | ||
NM_001205392.1 | NP_001192321.1 | 3 | 428 | |||
NM_029846.4 | NP_084122.2 | 3 | 428 | |||
GenBank mRNA | AB476646.1 | - | 3 | 239 | ||
AK030983.1 | - | 3 | 369 | |||
AK048873.1 | - | 3 | 347 | |||
AK131120.1 | - | 3 | 327 | |||
AK134333.1 | - | 3 | 384 | |||
AK145673.1 | - | 3 | 366 | |||
AK155358.1 | - | 3 | 364 | |||
AK163246.1 | - | 3 | 364 | |||
AK170073.1 | - | 3 | 362 | |||
BC049122.1 | - | 3 | 353 | 6813377 |