Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256115.1 | NP_001243044.1 | 3 | 301 | ||
NM_001256116.1 | NP_001243045.1 | 2 | 238 | |||
NM_001256117.1 | NP_001243046.1 | 3 | 301 | |||
NM_001256118.1 | NP_001243047.1 | 2 | 238 | |||
NM_024451.2 | NP_077771.1 | 3 | 301 | |||
GenBank mRNA | AK014585.1 | - | 3 | 299 | ||
AK036187.1 | - | 3 | 278 | |||
AK160281.1 | - | 3 | 259 | |||
AK161959.1 | - | 3 | 273 | |||
AK162533.1 | - | 3 | 272 | |||
AK167686.1 | - | 3 | 273 | |||
AK181458.1 | - | 2 | 172 | |||
AK196775.1 | - | 2 | 172 | |||
AK198340.1 | - | 2 | 172 | |||
AK200268.1 | - | 2 | 172 | |||
AY682989.1 | - | 2 | 218 | |||
BC047928.1 | - | 3 | 268 | 5321879 | ||
BC048156.1 | - | 3 | 270 | 5698402 | ||
BQ551727.1 | - | 2 | 242 | |||
BY036585.1 | - | 3 | 301 | |||
CK389844.1 | - | 3 | 301 | |||
HQ402597.1 | - | 2 | 218 |