Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029857.3 | NP_084133.1 | 11, 12 | 1639 | ||
GenBank mRNA | AK014571.1 | - | 10, 11 | 1240 | ||
AK029092.1 | - | 11, 12 | 1639 | |||
AK054337.1 | - | 10, 11 | 1468 | |||
AK170619.1 | - | 11, 12 | 1368 | |||
AK171189.1 | - | 11, 12 | 1385 | |||
BC013471.1 | - | 10, 11 | 1265 | 4221072 |